Läs mer
1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide, TRC
Produktkod. 30138461
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138461 250 mg
30003963 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM320535250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 139756-01-7
Molekylformel C8 H12 N4 O3
Molekylvikt (g/mol) 212.21
Synonym 1H-Pyrazole-5-carboxamide, 1-methyl-4-nitro-3-propyl-, 1-Methyl-4-nitro-3-propyl-1H-pyrazole-5-carboxamide, 1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
IUPAC-namn 2-methyl-4-nitro-5-propylpyrazole-3-carboxamide
LEDER CCCc1nn(C)c(C(=O)N)c1[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 1-Methyl-4-nitro-3-propylpyrazole-5-carboxamide
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C8H12N4O3/c1-3-4-5-6(12(14)15)7(8(9)13)11(2)10-5/h3-4H2,1-2H3,(H2,9,13)
Formel vikt 212.0909
Procent renhet >95%