Läs mer
1-Methyl-4-(propan-2-ylidene)cyclohexane-1,2-diol (Mixture of Diastereomers), TRC
Produktkod. 30475083
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30475083 100 mg
1 options
Produktkod. 30475083 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM908190100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methyl-4-(propan-2-ylidene)cyclohexane-1,2-diol (Mixture of Diastereomers)
CAS 89708-25-8
Molekylformel C10H18O2
Kvantitet 100 mg
Synonym 1-methyl-4-(propan-2-ylidene)cyclohexane-1,2-diol (Mixture of Diastereomers)
InChI formel InChI=1S/C10H18O2/c1-7(2)8-4-5-10(3,12)9(11)6-8/h9,11-12H,4-6H2,1-3H3
LEDER OC1CC(=C(C)C)CCC1(O)C
Molekylvikt (g/mol) 170.25