Läs mer
MedChemExpress 1-Methyl-7-nitroisatoic anhydride
Produktkod. 16468768
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
Förpackningsstorlek:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16468768 10 mg 10mg
16431198 5 mg 5mg
16444687 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16468768 Leverantör MedChemExpress Leverantörsnummer HYD0913
MedChemExpress 1-methyl-7-nitroisatoic anhydride (1M7) is a reagent that detects local nucleotide flexibility, for probing 2'-hydroxyl reactivity.
Specifikationer
Innehåll och lagring 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Produkttyp Dye Reagents
Kvantitet 10 mg
Frakt skick Room temperature
CAS 73043-80-8
Molekylvikt (g/mol) 222.15 g/mol
Synonym 1M7
Mål HBV
Löslighet DMSO : 50 mg/mL (225.07 mM; Need ultrasonic)