Läs mer
1-Methyl Adenosine, TRC
Produktkod. 30137458
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137458 100 mg
30135726 50 mg
30008497 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137458 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM285065100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15763-06-1
Molekylformel C11H15N5O4
Molekylvikt (g/mol) 281.27
Synonym (2R,3S,4R,5R)-2-(Hydroxymethyl)-5-(6-imino-1-methyl-1H-purin-9(6H)-yl)tetrahydrofuran-3,4-diol;
IUPAC-namn (2R,3S,4R,5R)-2-(hydroxymethyl)-5-(6-imino-1-methylpurin-9-yl)oxolane-3,4-diol
LEDER CN1C=NC2=C(C1=N)N=CN2C3C(C(C(O3)CO)O)O
Specifikationer
Kemiskt namn eller material 1-Methyl Adenosine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C11H15N5O4/c1-15-3-14-10-6(9(15)12)13-4-16(10)11-8(19)7(18)5(2-17)20-11/h3-5,7-8,11-12,17-19H,2H2,1H3/t5-,7-,8-,11-/m1/s1
Formel vikt 281.112
Procent renhet >95%