Läs mer
Tocris Bioscience™ 1-Methyl-D-tryptophan
GREENER_CHOICE
Indoleamine 2,3-dioxygenase (IDO) inhibitor
Brand:  Tocris Bioscience™ 5698/50
Encompass
Produktkod. 15787056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
1-Methyl-D-tryptophan is an indoleamine 2,3-dioxygenase (IDO) inhibitor (IC50 = 7 μM); disrupts tryptophan catabolism. Enhances the antitumor and antiviral immunoresponses of CD8+ T-cells in vitro. Reduces tumor volume in mice with xenografts overexpressing IDO.
Specifikationer
Specifikationer
110117-83-4
1MT