Learn More
1-Methyl-Inosine, TRC
Product Code. 30137645
Change view
Click to view available options
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30137645 1 g
30007343 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137645 Supplier Toronto Research Chemicals Supplier No. TRCM3136001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2140-73-0
Molekylformel C11H14N4O5
Molekylvikt (g/mol) 282.25
Synonym 1-Methylinosine, N1-Methylinosine;
IUPAC-namn 9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1-methylpurin-6-one
LEDER CN1C=NC2=C(C1=O)N=CN2C3C(C(C(O3)CO)O)O
Specifications
Kemiskt namn eller material 1-Methyl-Inosine
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C11H14N4O5/c1-14-3-13-9-6(10(14)19)12-4-15(9)11-8(18)7(17)5(2-16)20-11/h3-5,7-8,11,16-18H,2H2,1H3/t5-,7-,8-,11-/m1/s1
Formel vikt 282.096
Procent renhet >95%