Läs mer
1-Methyl-pyridinium Chloride, TRC
Produktkod. 30479340
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30479340 1 g
1 options
Produktkod. 30479340 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3280601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methyl-pyridinium Chloride
CAS 7680-73-1
Molekylformel C6H8ClN
Kvantitet 1 g
Synonym 1-Methylpyridinium Chloride, Methylpyridinium Chloride, N-Methylpyridinium Chloride
InChI formel InChI=1S/C6H8N.ClH/c1-7-5-3-2-4-6-7;/h2-6H,1H3;1H/q+1;/p-1
LEDER C[N+]1=CC=CC=C1.[Cl-]
Molekylvikt (g/mol) 129.59