Läs mer
1-Methyl Xanthine, TRC
Produktkod. 30135038
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135038 1 g
30133459 250 mg
17969994 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135038 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3385751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6136-37-4
Molekylformel C6 H6 N4 O2
Molekylvikt (g/mol) 166.14
Synonym 1H-Purine-2,6-dione, 3,7-dihydro-1-methyl- (9CI), Xanthine, 1-methyl- (6CI,7CI,8CI), 3,9-Dihydro-1-methyl-1H-purine-2,6-dione, 1-Methyl-2,3,6,9-tetrahydro-1H-purine-2,6-dione, 1-Methyl-3,7-dihydro-purine-2,6-dione, 1-Methylxanthine, 2-Hydroxy-1-methyl-6,9-dihydro-1H-purin-6-one, 1H-Purine-2,6-dione, 3,9-dihydro-1-methyl-
IUPAC-namn 1-methyl-3,9-dihydropurine-2,6-dione
LEDER CN1C(=O)Nc2[nH]cnc2C1=O
Specifikationer
Kemiskt namn eller material 1-Methylxanthine
Kvantitet 1 g
InChI formel InChI=1S/C6H6N4O2/c1-10-5(11)3-4(8-2-7-3)9-6(10)12/h2H,1H3,(H,7,8)(H,9,12)
Formel vikt 166.0491