missing translation for 'onlineSavingsMsg'
Läs mer

1-Methylcyclopentanol, TRC

Produktkod. 30468501
Change view
Click to view available options
Kvantitet:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30468501 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468501 Supplier Toronto Research Chemicals Supplier No. TRCM29526510G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 1-Methylcyclopentanol
CAS 1462-03-9
Molekylformel C6H12O
Kvantitet 10 g
Synonym 1-Hydroxy-1-methylcyclopentane, 1-Methyl-1-cyclopentanol, 1-Methyl-1-hydroxycyclopentane, 1-Methylcyclopentan-1-ol, 1-Methylcyclopentyl Alcohol, NSC 23230
InChI formel InChI=1S/C6H12O/c1-6(7)4-2-3-5-6/h7H,2-5H2,1H3
LEDER CC1(O)CCCC1
IUPAC-namn 1-methylcyclopentan-1-ol
Molekylvikt (g/mol) 100.16
Formel vikt 100.0888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.