Läs mer
1-Methylethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate, Mikromol™
Produktkod. 17981506
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981506 100 mg
17903256 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17981506 Leverantör Mikromol Leverantörsnummer MM0084.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Methylethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methylethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
CAS 76508-37-7
Analyt- eller komponentnamn 1-Methylethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate
Frakt skick Room Temperature
Molekylformel C12 H13 N O5 S
Certifikat och efterlevnad ISO 17025
Synonym 1-Methylethyl (1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetate, Piroxicam Imp. F (EP), Piroxicam Impurity F
InChI formel InChI=1S/C12H13NO5S/c1-8(2)18-11(14)7-13-12(15)9-5-3-4-6-10(9)19(13,16)17/h3-6,8H,7H2,1-2H3
LEDER CC(C)OC(=O)CN1C(=O)c2ccccc2S1(=O)=O
IUPAC-namn propan-2-yl 2-(1,1,3-trioxo-1,2-benzothiazol-2-yl)acetate
Molekylvikt (g/mol) 283.3
Kvantitet 100 mg
Formel vikt 283.0514 g/mol