Läs mer
1-Methylnicotinamide Iodide, Mikromol™
Produktkod. 30052425
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052425 25 mg
17952776 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052425 Leverantör Mikromol Leverantörsnummer MM0552.050025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Methylnicotinamide Iodide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methylnicotinamide Iodide
CAS 6456-44-6
Analyt- eller komponentnamn 1-Methylnicotinamide Iodide
Frakt skick Room Temperature
Molekylformel C7 H9 N2 O . I
Certifikat och efterlevnad ISO 17025
Synonym Pyridinium, 3-(aminocarbonyl)-1-methyl-, iodide (1:1), 3-Carbamoyl-1-methylpyridinium iodide (6CI,7CI), Pyridinium, 3-(aminocarbonyl)-1-methyl-, iodide (9CI), Pyridinium, 3-carbamoyl-1-methyl-, iodide (8CI), Pyridinium, 3-carbamyl-1-methyl-, iodide (4CI), 1-Methylnicotinamide iodide, Nicotinamide methiodide
InChI formel InChI=1S/C7H8N2O.HI/c1-9-4-2-3-6(5-9)7(8)10;/h2-5H,1H3,(H-,8,10);1H
LEDER [I-].C[n+]1cccc(c1)C(=O)N
IUPAC-namn 1-methylpyridin-1-ium-3-carboxamide;iodide
Molekylvikt (g/mol) 264.06
Kvantitet 25 mg
Formel vikt 263.976 g/mol