missing translation for 'onlineSavingsMsg'
Läs mer

1-Methylpyrrole-2-carbonyl chloride, 95%

Produktkod. p-7047266
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11364995 1 g 1g
11374995 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11364995 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L18431.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 26214-68-6 | C6H6ClNO | 143.57 g/mol

It is applied in the preparation of 5-(Pyrrolylcarbonyl)- and 5-(Imidazolylcarbonyl)pyrimidines and use also in the novel fluorescent, minor groove and sequence specific recognition element: design, synthesis, and DNA binding properties of p-Anisylbenzimidazole-imidazole/pyrrole-containing polyamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in the preparation of 5-(Pyrrolylcarbonyl)- and 5-(Imidazolylcarbonyl)pyrimidines and use also in the novel fluorescent, minor groove and sequence specific recognition element: design, synthesis, and DNA binding properties of p-Anisylbenzimidazole-imidazole/pyrrole-containing polyamides.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store away from water/moisture, bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Store in freezer at -20°C.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 26214-68-6
Molekylformel C6H6ClNO
Molekylvikt (g/mol) 143.57
MDL-nummer MFCD02094021
InChI-nyckel JOYDZQJINHJNPM-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carbonyl chloride, 1h-pyrrole-2-carbonyl chloride, 1-methyl, 1-methyl-1h-pyrrole-2-carboxylic acid chloride, acmc-1chjw, 1-methyl-2-pyrroyl chloride, n-methyl-2-pyrroloyl chloride, n-methylpyrrolecarbonyl chloride, n-methylpyrrole carbonyl chloride, 1-methylpyrrole-2-carbonylchloride, 1-methylpyrrole-2-carboyl chloride
PubChem CID 2795092
IUPAC-namn 1-methylpyrrole-2-carbonyl chloride
LEDER CN1C=CC=C1C(=O)Cl

Specifikationer

Kemiskt namn eller material 1-Methylpyrrole-2-carbonyl chloride
Kokpunkt 72°C to 74°C (10 mmHg)
Brytningsindex 1.57
Kvantitet 1 g
UN-nummer UN3265
Beilstein 472022
Känslighet Air and moisture sensitive
Löslighetsinformation Reacts with water.
Formel vikt 143.57
Procent renhet 95%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.