Läs mer
1-Methylthio-1-methylamino-2-nitroethylene, 97%
Produktkod. 11465197
Change view
Kvantitet:
10 g
50 g
250 g
Förpackningsstorlek:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11465197 250 g 250g
11445197 10 g 10g
11455197 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11465197 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B22608.30
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 61832-41-5 | C4H8N2O2S | 148.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 61832-41-5
Molekylformel C4H8N2O2S
Molekylvikt (g/mol) 148.18
MDL-nummer MFCD00040994
InChI-nyckel YQFHPXZGXNYYLD-ONEGZZNKSA-N
Synonym n-methyl-1-methylthio-2-nitroethenamine, unii-a9ls0uv4e3, ethenamine, n-methyl-1-methylthio-2-nitro, a9ls0uv4e3, n-methyl-1-methylthio-2-nitro-1-etheneamine, 1-methylamino-1-methylthio-2-nitroethylene, e-n-methyl-1-methylthio-2-nitroethenamine, e-n-methyl-1-methylsulfanyl-2-nitroethenamine
PubChem CID 3035401
IUPAC-namn (E)-N-methyl-1-methylsulfanyl-2-nitroethenamine
LEDER CNC(=C[N+](=O)[O-])SC
Specifikationer
Kemiskt namn eller material 1-Methylthio-1-methylamino-2-nitroethylene
Smältpunkt 109°C to 113°C
Kvantitet 250 g
Beilstein 2828017
Formel vikt 148.18
Procent renhet 97%