Läs mer
1-Naphthol 2,3,4-Tri-O-acetyl-Beta-D-glucuronide Methyl Ester, TRC
Produktkod. 30484002
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30484002 500 mg
30470404 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30484002 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN369000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 18404-55-2
Molekylformel C23 H24 O10
Molekylvikt (g/mol) 460.43
Synonym 1-Naphthol 2,3,4-Tri-O-Acetyl-B-D-Glucuronide Methyl Ester
IUPAC-namn methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-naphthalen-1-yloxyoxane-2-carboxylate
LEDER COC(=O)[C@H]1O[C@@H](Oc2cccc3ccccc23)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
Specifikationer
Kemiskt namn eller material 1-Naphthol 2,3,4-Tri-O-acetyl-Beta-D-glucuronide Methyl Ester
Kvantitet 500 mg
InChI formel InChI=1S/C23H24O10/c1-12(24)29-18-19(30-13(2)25)21(31-14(3)26)23(33-20(18)22(27)28-4)32-17-11-7-9-15-8-5-6-10-16(15)17/h5-11,18-21,23H,1-4H3/t18-,19-,20-,21+,23+/m0/s1
Formel vikt 460.1369