Learn More
1-Naphthol-D7, TRC
Product Code. 30480873
Change view
Click to view available options
Kvantitet:
50 mg
Product Code. Kvantitet
30480873 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30480873 Supplier Toronto Research Chemicals Supplier No. TRCN36799650MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 1-Naphthol-d7 (ring d7)
CAS 124251-84-9
Anteckningar om renhetsgrad HPLC
Molekylformel C10 2H7 H O
Kvantitet 50 mg
Synonym 1-Naphthalen-2,3,4,5,6,7,8-d7-ol, 1-Naphthol-d7, 1-Naphthol (ring-D7)
InChI formel InChI=1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H/i1D,2D,3D,4D,5D,6D,7D
LEDER [2H]c1c([2H])c([2H])c2c(O)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC-namn 2,3,4,5,6,7,8-heptadeuterionaphthalen-1-ol
Molekylvikt (g/mol) 151.21
Formel vikt 151.1015