Läs mer
1-Naphthylacetyl Spermine Trihydrochloride, TRC
Produktkod. 30481954
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481954 10 mg
30476412 250 mg
30468417 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30481954 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN37780010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1049731-36-3
Molekylformel C22H37Cl3N4O
Molekylvikt (g/mol) 479.91
Synonym N-[3-[[4-[(3-Aminopropyl)amino]butyl]amino]propyl]-1-naphthaleneacetamide Trihydrochloride, Naspm Trihydrochloride
IUPAC-namn N-[3-[4-(3-aminopropylamino)butylamino]propyl]-2-naphthalen-1-ylacetamide;trihydrochloride
LEDER NCCCNCCCCNCCCNC(CC1=CC=CC2=CC=CC=C21)=O.Cl.Cl.Cl
Specifikationer
Kemiskt namn eller material 1-Naphthylacetyl Spermine Trihydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C22H34N4O.3ClH/c23-12-6-15-24-13-3-4-14-25-16-7-17-26-22(27)18-20-10-5-9-19-8-1-2-11-21(19)20;;;/h1-2,5,8-11,24-25H,3-4,6-7,12-18,23H2,(H,26,27);3*1H
Formel vikt 478.2033