Läs mer
1-Naphthylamine, TRC
Produktkod. 30133204
Change view
Kvantitet:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133204 25 g
30136528 10 g
18072010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133204 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN37801525G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 134-32-7
Molekylformel C10 H9 N
Molekylvikt (g/mol) 143.19
Synonym 1-Naphthylamine (8CI), 1-Aminonaphthalene, 1-Naphthalamine, C.I. 37265, C.I. Azoic Diazo Component 114, Fast Garnet Base B, Naphthalen-1-ylamine, Naphthalidam, Naphthalidine, α-Aminonaphthalene, α-Naphthylamine
IUPAC-namn naphthalen-1-amine
LEDER Nc1cccc2ccccc12
Specifikationer
Kemiskt namn eller material 1-Aminonaphthalene
Kvantitet 25 g
InChI formel InChI=1S/C10H9N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,11H2
Formel vikt 143.0735