Läs mer
1-Naphthylhydrazine, TRC
Produktkod. 30474433
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474433 100 mg
30475920 10 mg
30472667 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474433 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN368528100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2243-55-2
Molekylformel C10H10N2
Molekylvikt (g/mol) 158.2
IUPAC-namn naphthalen-1-ylhydrazine
LEDER NNc1cccc2ccccc12
Specifikationer
Kemiskt namn eller material 1-Naphthylhydrazine
Kvantitet 100 mg
InChI formel InChI=1S/C10H10N2/c11-12-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,12H,11H2
Formel vikt 158.0844