missing translation for 'onlineSavingsMsg'
Learn More

1-Nitro-2,5-bis(trifluoromethyl)benzene, 98%, Thermo Scientific™

Catalog Number p-7055630
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
320-88-7
C8H3F6NO2
259.107
MFCD00236674
BZKVUOHHNCMTLH-UHFFFAOYSA-N
2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
2734048
2-nitro-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F
This item is not returnable. View return policy
320-88-7
C8H3F6NO2
259.107
MFCD00236674
BZKVUOHHNCMTLH-UHFFFAOYSA-N
2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
2734048
2-nitro-1,4-bis(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.570 g/mL
Boiling Point 90°C (15 mmHg)
Flash Point 76°C (169°F)
Refractive Index 1.427
Quantity 5 g
Beilstein 2508751
Sensitivity Light sensitive
Formula Weight 259.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Nitro-2,5-bis(trifluoromethyl)benzene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'