Läs mer
1-Nitropiperazine, TRC
Produktkod. 30480564
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480564 100 mg
30469231 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30480564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN518290100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 42499-41-2
Molekylformel C4H9N3O2
Molekylvikt (g/mol) 131.13
Synonym N-Nitropiperazine, 1-Nitropiperazine, NSC 243817
IUPAC-namn 1-nitropiperazine
LEDER [O-][N+](=O)N1CCNCC1
Specifikationer
Kemiskt namn eller material 1-Nitropiperazine
Kvantitet 100 mg
InChI formel InChI=1S/C4H9N3O2/c8-7(9)6-3-1-5-2-4-6/h5H,1-4H2
Formel vikt 131.0695