missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nitropropane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
500 g
Description
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.
Solubility
Soluble in water. (1.4 g/L)
Notes
Store in cool place. Keep away from oxidizing agents.
The solvent of 1-Nitropropane is mainly used coatings, inks and separation processes.
Solubility
Soluble in water. (1.4 g/L)
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Melting Point | -108°C |
Boiling Point | 131°C to 132°C |
Flash Point | 34°C (93°F) |
Odor | Unpleasant |
Linear Formula | CH3CH2CH2NO2 |
Quantity | 100 g |
UN Number | UN2608 |
Beilstein | 506236 |
Merck Index | 14,6627 |
Formula Weight | 89.09 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'