Läs mer
1-Nitrosopyrrolidine, TRC
Produktkod. 30005537
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005537 5 g
30012227 10 g
17973395 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005537 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5459505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 930-55-2
Molekylformel C4 H8 N2 O
Molekylvikt (g/mol) 100.12
Synonym Pyrrolidine, 1-nitroso-, 1-Nitrosopyrrolidine, N-Nitrosopyrrolidine, NPYR, NSC 18797
IUPAC-namn 1-nitrosopyrrolidine
LEDER O=NN1CCCC1
Specifikationer
Kemiskt namn eller material N-Nitrosopyrrolidine
Anteckningar om renhetsgrad GC
Kvantitet 5 g
InChI formel InChI=1S/C4H8N2O/c7-5-6-3-1-2-4-6/h1-4H2
Formel vikt 100.0637
Procent renhet >95%