Läs mer
1-O-(4,4'-Dimethoxytrityl)-2-aminopropan-3-ol, TRC
Produktkod. 30375656
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30375656 100 mg
30369931 1 g
30373123 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30375656 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD451896100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 154928-41-3
Molekylformel C24H27NO4
Molekylvikt (g/mol) 393.48
Synonym 2-Amino-3-[bis(4-methoxyphenyl)phenylmethoxy]-1-propanol
LEDER COc1ccc(cc1)C(OCC(N)CO)(c2ccccc2)c3ccc(OC)cc3
Specifikationer
Kemiskt namn eller material 1-O-(4,4'-Dimethoxytrityl)-2-aminopropan-3-ol
Kvantitet 100 mg
InChI formel InChI=1S/C24H27NO4/c1-27-22-12-8-19(9-13-22)24(29-17-21(25)16-26,18-6-4-3-5-7-18)20-10-14-23(28-2)15-11-20/h3-15,21,26H,16-17,25H2,1-2H3
Formel vikt 393.194