Learn More
1-Octacosanol, TRC
Product Code. 30010278
Change view
Click to view available options
Quantity:
500 mg
2.5 g
25 g
Unit Size:
2.5g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010278 25 g 25g
30005324 2.5 g 2.5g
17913215 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010278 Supplier Toronto Research Chemicals Supplier No. TRCO23965025G
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 557-61-9
Molecular Formula C28 H58 O
Molecular Weight (g/mol) 410.76
Synonym Cluytyl alcohol, Montanyl alcohol, NSC 10770, NSC 57768, Octacosanol, Octacosyl alcohol, n-Octacosanol, 1-Octacosanol
IUPAC Name octacosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCO
Specifications
Quantity 25 g
InChI Formula InChI=1S/C28H58O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29/h29H,2-28H2,1H3
Formula Weight 410.4488
Chemical Name or Material 1-Octacosanol