missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecyne, 96%, Thermo Scientific Chemicals

Catalog Number p-7031985
Click to view available options
Quantity:
1 g
10 g
629-89-0
C18H34
250.47
MFCD00015088
IYDNQWWOZQLMRH-UHFFFAOYSA-N
1-octadecyne, octadecyne, octadec-1-in, cetylacetylene, hexadecylacetylene, acmc-1b2rb, qspl 036
69425
octadec-1-yne
CCCCCCCCCCCCCCCCC#C
629-89-0
C18H34
250.47
MFCD00015088
IYDNQWWOZQLMRH-UHFFFAOYSA-N
1-octadecyne, octadecyne, octadec-1-in, cetylacetylene, hexadecylacetylene, acmc-1b2rb, qspl 036
69425
octadec-1-yne
CCCCCCCCCCCCCCCCC#C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 21°C to 24°C
Density 0.803
Boiling Point 313°C
Quantity 1 g
Formula Weight 250.47
Percent Purity ≥97%
Chemical Name or Material 1-Octadecyne

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'