missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Octyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
(1-Octyl)triphenylphosphonium bromide is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. Store under dry inert gas. It is hygroscopic in nature.
Specifications
Melting Point | 61°C to 63°C |
Linear Formula | CH3(CH2)7P(C6H5)3Br |
Quantity | 5 g |
Beilstein | 3642460 |
Sensitivity | Hygroscopic |
Formula Weight | 455.42 |
Percent Purity | 97% |
Chemical Name or Material | (1-Octyl)triphenylphosphonium bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'