Läs mer
1-Oxo Fexofenadine Methyl Ester, TRC
Produktkod. 30411231
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30411231 25 g
1 options
Produktkod. 30411231 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO87004025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Oxo Fexofenadine Methyl Ester
CAS 154477-55-1
Molekylformel C33H39NO4
Kvantitet 25 g
Synonym 4-[4-[4-(Hydroxydiphenylmethyl)-1-piperidinyl]-1-oxobutyl]-alpha,alpha-dimethylbenzeneacetic Acid Methyl Ester, 2-[4-[4-[4-(Hydroxydiphenylmethyl)-1-piperidinyl]-1-oxobutyl]phenyl]-2,2-dimethylacetic Acid Methyl Ester, Methyl 4-[4-[4-(Hydroxydiphenylmethyl)-1-piperidinyl]-1-oxobutyl]-alpha,alpha-dimethylphenylacetate
InChI formel InChI=1S/C33H39NO4/c1-32(2,31(36)38-3)26-18-16-25(17-19-26)30(35)15-10-22-34-23-20-29(21-24-34)33(37,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29,37H,10,15,20-24H2,1-3H3
LEDER COC(=O)C(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
IUPAC-namn methyl 2-[4-[4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butanoyl]phenyl]-2-methylpropanoate
Molekylvikt (g/mol) 513.67
Formel vikt 513.2879