Läs mer
1-Oxo Mirtazapine (Mirtazapine Impurity C), TRC
Produktkod. 30418754
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30418754 5 mg
1 options
Produktkod. 30418754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8579905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Oxo Mirtazapine
CAS 191546-96-0
Molekylformel C17 H17 N3 O
Kvantitet 5 mg
Synonym Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepin-1(2H)-one','3,4,10,14b-tetrahydro-2-methyl-, Mirtazapine Related Compound C (USP)
InChI formel InChI=1S/C17H17N3O/c1-19-9-10-20-15(17(19)21)14-7-3-2-5-12(14)11-13-6-4-8-18-16(13)20/h2-8,15H,9-11H2,1H3
LEDER CN1CCN2C(C1=O)c3ccccc3Cc4cccnc24
Molekylvikt (g/mol) 279.34
Formel vikt 279.1372