Läs mer
1-Oxyl-2,2,5,5-tetramethyl-∆3-(methanesulfonyloxymethyl)pyrroline, TRC
Produktkod. 30428318
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428318 10 mg
30427147 25 mg
30416540 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428318 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO87240010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76893-27-1
Molekylformel C10 H18 N O4 S
Molekylvikt (g/mol) 248.32
IUPAC-namn (1-λ^{1}-oxidanyl-2,2,5,5-tetramethylpyrrol-3-yl)methyl methanesulfonate
LEDER CC1(C)C=C(COS(=O)(=O)C)C(C)(C)N1[O]
Specifikationer
Kemiskt namn eller material 1-Oxyl-2,2,5,5-tetramethyl-delta3-(methanesulfonyloxymethyl)pyrroline
Kvantitet 10 mg
InChI formel InChI=1S/C10H18NO4S/c1-9(2)6-8(7-15-16(5,13)14)10(3,4)11(9)12/h6H,7H2,1-5H3
Formel vikt 248.0957