Läs mer
(1-Oxyl-2,2,5,5-tetramethylpyrrolidin-3-yl) Methyl Methanethiosulfonate, TRC
Produktkod. 30409617
Change view
Kvantitet:
2.5 mg
10 mg
50 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30409617 50 mg -
30426419 10 mg -
30422306 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30409617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO87390050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201403-46-5
Molekylformel C10 H20 N O3 S2
Molekylvikt (g/mol) 266.4
IUPAC-namn 1-λ^{1}-oxidanyl-2,2,5,5-tetramethyl-3-(methylsulfonylsulfanylmethyl)pyrrolidine
LEDER CC1(C)CC(CSS(=O)(=O)C)C(C)(C)N1[O]
Specifikationer
Kemiskt namn eller material (1-Oxyl-2,2,5,5-tetramethylpyrrolidin-3-yl) Methyl Methaneth
Kvantitet 50 mg
InChI formel InChI=1S/C10H20NO3S2/c1-9(2)6-8(7-15-16(5,13)14)10(3,4)11(9)12/h8H,6-7H2,1-5H3
Formel vikt 266.0885