Läs mer
1-Palmitoyl-2-oleoyl-3-stearoyl-rac-glycerol, TRC
Produktkod. 30135650
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135650 100 mg
17968414 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135650 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP162100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2190-27-4
Molekylformel C55 H104 O6
Molekylvikt (g/mol) 861.41
Synonym (Z)-9-Octadecenoic Acid 1-[[(1-Oxohexadecyl)oxy]methyl]-2-[(1-oxooctadecyl)oxy]ethyl Ester, 1-Palmito-2-oleo-3-stearin, 1-Palmito-2-oleostearin, 1-Palmitoyl-2-oleoyl-3-stearin, 1-Palmitoyl-2-oleoyl-3-stearoylglycerol, 1-Palmityl-2-oleyl-3-stearylglycerol, 1-Stearo-2-palmito-3-olein, 1-Stearoyl-2-oleoyl-3-palmitoylglycerol, 2-(Oleoyloxy)-1-(palmitoyloxy)-3-(stearoyloxy)propane, 2-Oleo-3-palmito-1-stearin, 2-Oleo-3-stearopalmitin, Glyceryl-1-palmitate-2-oleate-3-stearate, POS;
IUPAC-namn [3-hexadecanoyloxy-2-[(Z)-octadec-9-enoyl]oxypropyl] octadecanoate
LEDER CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCC)OC(=O)CCCCCCC\C=C/CCCCCCCC
Specifikationer
Kemiskt namn eller material 1-Palmitoyl-2-oleoyl-3-stearoyl-rac-glycerol
Kvantitet 100 mg
InChI formel InChI=1S/C55H104O6/c1-4-7-10-13-16-19-22-25-27-30-33-36-39-42-45-48-54(57)60-51-52(50-59-53(56)47-44-41-38-35-32-29-24-21-18-15-12-9-6-3)61-55(58)49-46-43-40-37-34-31-28-26-23-20-17-14-11-8-5-2/h26,28,52H,4-25,27,29-51H2,1-3H3/b28-26-
Formel vikt 860.7833