Läs mer
1-Palmitoyl-2-oleoyl-rac-glycerol, TRC
Produktkod. 30010778
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010778 25 mg
30007518 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010778 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP16095025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3123-73-7
Molekylformel C37 H70 O5
Molekylvikt (g/mol) 594.95
IUPAC-namn (1-hexadecanoyloxy-3-hydroxypropan-2-yl) (Z)-octadec-9-enoate
LEDER CCCCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCC\C=C/CCCCCCCC
Specifikationer
Kemiskt namn eller material 1-Palmitoyl-2-oleoyl-rac-glycerol
Kvantitet 25 mg
InChI formel InChI=1S/C37H70O5/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(40)42-35(33-38)34-41-36(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h17-18,35,38H,3-16,19-34H2,1-2H3/b18-17-
Formel vikt 594.5223