Läs mer
1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine, TRC
Produktkod. 30420466
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420466 10 mg
30414285 100 mg
30421463 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP16100010MG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 26853-31-6
Molekylformel C42H82NO8P
Molekylvikt (g/mol) 760.08
Synonym 3,5,8-Trioxa-4-phosphahexacos-17-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-, inner salt, 4-oxide, (7R,17Z)-, 3,5,8-Trioxa-4-phosphahexacos-17-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-, hydroxide, inner salt, 4-oxide, [R-(Z)]-, Choline, hydroxide, dihydrogen phosphate, inner salt, ester with 1-palmito-2-olein, L- (8CI), 1-Hexadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-3-sn-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-3-sn-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-L-α-lecithin, 1-Palmitoyl-2-oleoyl-L-α-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphorylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycero-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphatidylcholine, 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine, 1-Palmitoyl-2-oleoyl-sn-glyceryl-3-phosphorylcholine, 1-Palmitoyl-2-oleoylphosphatidylcholine, 1-Palmitoyl-2-oleyl-3-sn-phosphatidylcholine, 2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, 3,5,8-Trioxa-4-phosphahexacos-17-en-1-aminium, 4-hydroxy-N,N,N-trimethyl-9-oxo-7-[[(1-oxohexadecyl)oxy]methyl]-, inner salt, 4-oxide, [R-(Z)]-, L-1-Palmitoyl-2-oleoylphosphatidylcholine, L-Palmitoyloleoylphosphatidylcholine, L-α-1-Palmitoyl-2-oleoylphosphatidylcholine, POPC, Palmitoyloleoylphosphatidylcholine, β-Oleoyl-γ-palmitoyl-L-α-phosphatidylcholine
IUPAC-namn [(2R)-3-hexadecanoyloxy-2-[(Z)-octadec-9-enoyl]oxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
LEDER CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCC\C=C/CCCCCCCC
Specifikationer
Kemiskt namn eller material 1-Palmitoyl-2-oleoyl-sn-glycerol-3-phosphocholine
Kvantitet 10 mg
InChI formel InChI=1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1
Formel vikt 759.5778