Läs mer
1-Phenanthrenecarboxylic Acid, TRC
Produktkod. 30426494
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426494 10 mg
30411461 100 mg
30419167 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP29479510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 27875-89-4
Molekylformel C15H10O2
Molekylvikt (g/mol) 222.24
Synonym NSC 139679
IUPAC-namn phenanthrene-1-carboxylic acid
LEDER OC(=O)c1cccc2c1ccc3ccccc23
Specifikationer
Kemiskt namn eller material 1-Phenanthrenecarboxylic Acid
Kvantitet 10 mg
InChI formel InChI=1S/C15H10O2/c16-15(17)14-7-3-6-12-11-5-2-1-4-10(11)8-9-13(12)14/h1-9H,(H,16,17)
Formel vikt 222.0681