Läs mer
1-Phenyl-1,2-propanediol (Mixture of Diastereomers), TRC
Produktkod. 30134416
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134416 250 mg
30011247 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134416 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP336095250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1855-09-0
Molekylformel C9H12O2
Molekylvikt (g/mol) 152.19
Synonym (RS,RS)-2-Methyl-1-phenyl-1,2-ethanediol, 1,2-Dihydroxy-1-phenylpropane, 1-Phenyl-1,2-dihydroxypropane
IUPAC-namn 1-phenylpropane-1,2-diol
LEDER OC(C1=CC=CC=C1)C(O)C
Specifikationer
Kemiskt namn eller material 1-Phenyl-1,2-propanediol (Mixture of Diastereomers)
Kvantitet 250 mg
InChI formel InChI=1S/C9H12O2/c1-7(10)9(11)8-5-3-2-4-6-8/h2-7,9-11H,1H3
Formel vikt 152.0837