missing translation for 'onlineSavingsMsg'
Läs mer

[(1-Phenyl-1H-tetrazol-5-yl)sulfonyl]acetic Acid Ethyl Ester, TRC

Produktkod. 30419195
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419195 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30419195 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP337020100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material [(1-Phenyl-1H-tetrazol-5-yl)sulfonyl]acetic Acid Ethyl Ester
CAS 852657-49-9
Molekylformel C22 H22 O12
Kvantitet 100 mg
InChI formel InChI=1S/C22H22O12/c1-31-13-3-2-8(4-10(13)23)14-7-12(25)16-11(24)5-9(6-15(16)33-14)32-22-19(28)17(26)18(27)20(34-22)21(29)30/h2-6,14,17-20,22-24,26-28H,7H2,1H3,(H,29,30)/t14-,17?,18-,19?,20?,22-/m1/s1
LEDER COc1ccc(cc1O)[C@H]2CC(=O)c3c(O)cc(O[C@@H]4OC([C@H](O)C(O)C4O)C(=O)O)cc3O2
IUPAC-namn (3R,6S)-3,4,5-trihydroxy-6-[[(2R)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-4-oxo-2,3-dihydrochromen-7-yl]oxy]oxane-2-carboxylic acid
Molekylvikt (g/mol) 478.403
Formel vikt 478.111
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.