Läs mer
1-Phenyl-2-(4-N-Cbz-phenylamino-piperidin-1-yl)-propan-1-one, TRC
Produktkod. 30428638
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428638 50 mg
30411655 100 mg
30418859 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428638 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP33694850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 936498-12-3
Molekylformel C28H30N2O3
Molekylvikt (g/mol) 442.56
IUPAC-namn benzyl N-[1-(1-oxo-1-phenylpropan-2-yl)piperidin-4-yl]-N-phenylcarbamate
LEDER CC(N1CCC(CC1)N(C(=O)OCc2ccccc2)c3ccccc3)C(=O)c4ccccc4
Specifikationer
Kemiskt namn eller material 1-Phenyl-2-(4-N-Cbz-phenylamino-piperidin-1-yl)-propan-1-one
Kvantitet 50 mg
InChI formel InChI=1S/C28H30N2O3/c1-22(27(31)24-13-7-3-8-14-24)29-19-17-26(18-20-29)30(25-15-9-4-10-16-25)28(32)33-21-23-11-5-2-6-12-23/h2-16,22,26H,17-21H2,1H3
Formel vikt 442.2256