missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Phenyl-3-[2-(trifluoromethyl)phenyl]urea, 95%

Brand:  Alfa Aesar™ H61501.03

Additional Details : CAS Number : 895-40-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15468058

  • 7115.42 SEK / 1g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

1-Phenyl-3-[2-(trifluoromethyl)phenyl]urea
895-40-9
MFCD00424422
3383874

Specifications

1-Phenyl-3-[2-(trifluoromethyl)phenyl]urea
White
895-40-9
95%
MFCD00424422
KPQRZAREDAWPGZ-UHFFFAOYSA-N
280.25
280.25
1g
Solid
C14H11F3N2O
3383874
C1=CC=C(C=C1)NC(=O)NC2=CC=CC=C2C(F)(F)F
728635
Videos
SDS