Läs mer
1-Phenyl-3-azabicyclo[3.1.0]hexan-2-one, TRC
Produktkod. 30417281
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417281 100 mg
30427608 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30417281 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP320810100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 153275-06-0
Molekylformel C11H11NO
Molekylvikt (g/mol) 173.21
Synonym 1-Phenyl-3-aza-bicyclo[3.1.0]hexan-2-one
IUPAC-namn 1-phenyl-3-azabicyclo[3.1.0]hexan-2-one
LEDER O=C1NCC2CC12c3ccccc3
Specifikationer
Kemiskt namn eller material 1-Phenyl-3-azabicyclo[3.1.0]hexan-2-one
Kvantitet 100 mg
InChI formel InChI=1S/C11H11NO/c13-10-11(6-9(11)7-12-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,13)
Formel vikt 173.0841