Läs mer
1-fenyl-3-(piperidin-1-yl) propan-1-ett hydroklorid, Mikromol™
Beskrivning
1-fenyl-3-(piperidin-1-yl)propan-1-ett hydroklorid
Specifikationer
Specifikationer
| Kemiskt namn eller material | 1-Phenyl-3-(piperidin-1-yl)propan-1-one Hydrochloride |
| CAS | 886-06-6 |
| Analyt- eller komponentnamn | 1-Phenyl-3-(piperidin-1-yl)propan-1-one Hydrochloride |
| Frakt skick | Rumstemperatur |
| Molekylformel | C14 H19 N O . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Trihexyphenidyl Hydrochloride Imp. A (EP), Trihexyphenidyl Imp. A (EP), Trihexyphenidyl USP Related Compound A, Trihexyphenidyl USP RC A, 1-Phenyl-3-(piperidin-1-yl)propan-1-one Hydrochloride, Trihexyphenidyl Hydrochloride Imp. A (EP) as Hydrochloride, Piperidyl Propiophenone Hydrochloride, Pridinol Hydrochloride Imp. A as Hydrochloride, Trihexyphenidyl Related Compound A as Hydrochloride, Trihexyphenidyl Hydrochloride Impurity A as Hydrochloride, Trihexyphenidyl Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C14H19NO.ClH/c16-14(13-7-3-1-4-8-13)9-12-15-10-5-2-6-11-15;/h1,3-4,7-8H,2,5-6,9-12H2;1H |
| LEDER | Cl.O=C(CCN1CCCCC1)c2ccccc2 |
| IUPAC-namn | 1-fenyl-3-pipridin-1-ylpropan-1-one; hydroklorid |
| Visa mer |