Läs mer
1-Phenyl-N-propylpentan-2-amine Hydrochloride, TRC
Produktkod. 30133085
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133085 500 mg
30136512 1 g
30008584 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133085 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP321197500MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 119486-01-0
Molekylformel C14 H23 N . H Cl
Molekylvikt (g/mol) 241.8
Synonym N,alpha-Dipropylbenzeneethanamine Hydrochloride, (1-Phenylpentan-2-yl)(propyl)amine Hydrochloride
IUPAC-namn 1-phenyl-N-propyl-pentan-2-amine;hydrochloride
LEDER Cl.CCCNC(CCC)Cc1ccccc1
Specifikationer
Kemiskt namn eller material 1-Phenyl-N-propylpentan-2-amine hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C14H23N.ClH/c1-3-8-14(15-11-4-2)12-13-9-6-5-7-10-13;/h5-7,9-10,14-15H,3-4,8,11-12H2,1-2H3;1H
Formel vikt 241.1597