missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyldodecane, 97%, Thermo Scientific Chemicals

Catalog Number p-3779421
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
123-01-3
C18H30
246.43
MFCD00008974
KWKXNDCHNDYVRT-UHFFFAOYSA-N
1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
31237
dodecylbenzene
CCCCCCCCCCCCC1=CC=CC=C1
This item is not returnable. View return policy
123-01-3
C18H30
246.43
MFCD00008974
KWKXNDCHNDYVRT-UHFFFAOYSA-N
1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
31237
dodecylbenzene
CCCCCCCCCCCCC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -7°C
Density 0.8500g/mL
Boiling Point 288°C to 300°C
Flash Point 124°C
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5(CH2)11CH3
Refractive Index 1.4810 to 1.483
Quantity 10 g
Specific Gravity 0.85
Solubility Information Solubility in water: immiscible with water
Formula Weight 246.43
Percent Purity 97%
Chemical Name or Material 1-Phenyldodecane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'