Läs mer
1-(Phenylsulfonyl)pyrrole, 98%
Produktkod. 11404683
Change view
Kvantitet:
1 g
5 g
25 g
Förpackningsstorlek:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11404683 1 g 1g
11414683 5 g 5g
11424683 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11404683 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A11599.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 16851-82-4 | C10H9NO2S | 207.25 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16851-82-4
Molekylformel C10H9NO2S
Molekylvikt (g/mol) 207.25
MDL-nummer MFCD00067739
InChI-nyckel PPPXRIUHKCOOMU-UHFFFAOYSA-N
PubChem CID 140146
IUPAC-namn 1-(benzenesulfonyl)pyrrole
LEDER O=S(=O)(N1C=CC=C1)C1=CC=CC=C1
Specifikationer
Kemiskt namn eller material 1- (Phenylsulfonyl)pyrrole
Smältpunkt 87°C to 90°C
Kvantitet 1 g
Beilstein 1619427
Synonym 1-phenylsulfonyl pyrrole, 1-phenylsulfonyl-1h-pyrrole, n-benzenesulphonylpyrrole, 1-benzenesulfonyl pyrrole, n-benzenesulfonylpyrrole, 1-benzenesulfonyl-1h-pyrrole, n-phenylsulfonylpyrrole, phenylsulfonyl pyrrole, 1h-pyrrole,1-phenylsulfonyl, 1h-pyrrole, 1-phenylsulfonyl
Formel vikt 207.25
Procent renhet 98%