missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylvinylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7035499
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
143825-84-7
C14H19BO2
230.114
MFCD06659923
RMGBWPMWUZSIMH-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-1-phenylvinyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl, 1-phenylvinylboronic acid pinacolester, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
11031671
4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C(=C)C2=CC=CC=C2
This item is not returnable. View return policy
143825-84-7
C14H19BO2
230.114
MFCD06659923
RMGBWPMWUZSIMH-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-1-phenylvinyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid pinacol ester, 2-1-phenylethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl-1,3,2-dioxaborolane, 1-phenylvinylboronic acid, pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-phenylethenyl, 1-phenylvinylboronic acid pinacolester, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-1-phenylvinyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
11031671
4,4,5,5-tetramethyl-2-(1-phenylethenyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C(=C)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 40°C to 42°C
Quantity 250 mg
Formula Weight 230.11
Percent Purity 95%
Chemical Name or Material 1-Phenylvinylboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'