missing translation for 'onlineSavingsMsg'
Learn More

1-Piperazinecarboxaldehyde, 90%, Thermo Scientific Chemicals

Catalog Number p-3211419
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7755-92-2
C5H11N2O
115.16
MFCD00005963
MSSDTZLYNMFTKN-UHFFFAOYSA-O
1-formylpiperazine, 1-piperazinecarboxaldehyde, 1-piperazinecarbaldehyde, formylpiperazine, n-formylpiperazine, 1-formyl piperazine, piperazinecarbaldehyde, l-formylpiperazine, 4-formylpiperazine, n-formyl piperazine
82191
piperazine-1-carbaldehyde
O=CN1CC[NH2+]CC1
This item is not returnable. View return policy
7755-92-2
C5H11N2O
115.16
MFCD00005963
MSSDTZLYNMFTKN-UHFFFAOYSA-O
1-formylpiperazine, 1-piperazinecarboxaldehyde, 1-piperazinecarbaldehyde, formylpiperazine, n-formylpiperazine, 1-formyl piperazine, piperazinecarbaldehyde, l-formylpiperazine, 4-formylpiperazine, n-formyl piperazine
82191
piperazine-1-carbaldehyde
O=CN1CC[NH2+]CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 105.0°C to 107.0°C (1.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5097 to 1.5117
Quantity 25 g
Specific Gravity 1.1
Formula Weight 114.15
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-Piperazinecarboxaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Piperazinecarboxaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'