Läs mer
1-Piperideine, TRC
Produktkod. 30427738
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427738 10 mg
30409116 100 mg
30410210 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427738 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP99461810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 505-18-0
Molekylformel C5 H9 N
Molekylvikt (g/mol) 83.132
Synonym 2,3,4,5-Tetrahydropyridine, 3,4,5,6-Tetrahydropyridine, δ1-Piperideine
IUPAC-namn 2,3,4,5-tetrahydropyridine
LEDER C1CCN=CC1
Specifikationer
Kemiskt namn eller material 1-Piperideine
Kvantitet 10 mg
InChI formel InChI=1S/C5H9N/c1-2-4-6-5-3-1/h4H,1-3,5H2
Formel vikt 83.073