missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Propanol, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
Specifications
Formula Weight | 60.1 |
Grade | BAKER ANALYZED™ Reagent |
Quantity | 4 L |
Flash Point | 23.00°C |
Packaging | Narrow Mouth PVC-coated Amber Bottle |
Specific Gravity | 1L = 0.80kg |
Chemical Name or Material | 1-Propanol |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'