Läs mer
1-Pyrenemethylamine Hydrochloride, TRC
Produktkod. 30417934
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30417934 100 mg
1 options
Produktkod. 30417934 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP989505100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Pyrenemethylamine Hydrochloride
CAS 93324-65-3
Molekylformel C17H13N.HCl
Kvantitet 100 mg
Synonym 1-Aminomethylpyrene Hydrochloride, 1-Pyrenemethylammonium Chloride, 1-Pyrenylmethylamine Hydrochloride, 1-Pyrenemethanamine Hydrochloride
InChI formel InChI=1S/C17H13N.ClH/c18-10-14-7-6-13-5-4-11-2-1-3-12-8-9-15(14)17(13)16(11)12;/h1-9H,10,18H2;1H
LEDER Cl.NCc1ccc2ccc3cccc4ccc1c2c34
IUPAC-namn pyren-1-ylmethanamine;hydrochloride
Molekylvikt (g/mol) 267.8
Formel vikt 267.0815