missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(tert-Butoxycarbonyl)amino]cyclopropanecarboxylic acid, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Formula Weight | 201.22 |
Percent Purity | 97% |
Quantity | 250 mg |
Chemical Name or Material | 1-[(Tert-butoxycarbonyl)amino]cyclopropanecarboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'