Läs mer
1-tert-butyl-1H-pyrrole-3-carboxylic Acid, TRC
Produktkod. 30427175
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427175 500 mg
30418001 100 mg
30421525 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427175 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT136020500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1266364-46-8
Molekylformel C9H13NO2
Molekylvikt (g/mol) 167.21
Synonym 1-(1,1-Dimethylethyl)-1H-pyrrole-3-carboxylic Acid
IUPAC-namn 1-tert-butylpyrrole-3-carboxylic acid
LEDER CC(C)(C)n1ccc(c1)C(=O)O
Specifikationer
Kemiskt namn eller material 1-tert-butyl-1H-pyrrole-3-carboxylic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C9H13NO2/c1-9(2,3)10-5-4-7(6-10)8(11)12/h4-6H,1-3H3,(H,11,12)
Formel vikt 167.0946